Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

44318.09 44318.18
44318.09EA 218 EUR
44318.09 44318.18
Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)
Manganese(II) perchlorate hexahydrate

Purity: 99.995% CAS No.: 15364-94-0 Molecular Formula: Mn(ClO4)2.6H2O Formula Weight: 361.93 (253.84anhy) Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 1481 MDL No.: MFCD00150255

Kaava: Cl₂MnO₈·6H₂O
MW: 361.931 g/mol
Sulamispiste: 165 °C. (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150255
CAS-numero: 15364-94-0
EINECS: 237-390-9
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR