Magnetic stirring bars, PTFE, Micro, Spinbar®, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F371190008 F371190127
442-9112EA 19.9 EUR
442-9112 BELAF371190127
Magnetic stirring bars, PTFE, Micro, Spinbar®, SP Bel-Art
Sekoittajat Magneettisauva tai namu
PTFE coated micro magnetic stirring bars which are designed for stirring small volumes in vessels such as vials, tubes and gradient makers. Autoclave safe.