Nikkeli Kromi metalliseos (80:20 w%), lanka, Ø 0.05 mm (0.002 in)

Toimittaja: Thermo Fisher Scientific
Varoitus

44187.H9 44187.J5
44187.H9EA 586 EUR
44187.H9 44187.J5
Nikkeli Kromi metalliseos (80:20 w%), lanka, Ø 0.05 mm (0.002 in)
Nikkeli Kromi metalliseos (80:20 w%)

CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: ≈0.017g/m Odour: Odourless MDL No.: MFCD00673110

Storage Temperature: Huoneenlämpö
MDL numero: MFCD00673110
CAS-numero: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR