Electronic overhead stirrers, MICROSTAR control
Toimittaja: IKA
25001984 25001986 25001987
441-0464EA 1530 EUR
441-0464 441-0465 441-0466
Electronic overhead stirrers, MICROSTAR control
Sekoittajat Sekoitusmoottorit
These overhead stirrers are ideal for special applications. Combining high performance with excellence, the units require minimum space.
  • Fast, responsive display for maximum visibility and chemical resistance
  • Clear display shows all essential information at a glance
  • Integrated timer/counter for the control of kinetic sensitive reactions and reminders
  • USB interface for documenting parameters using labworldsoft® or installing firmware updates
  • Intuitive and simple handling; touch-sensitive surface for long service life
  • Temperature reading is shown on display