Kiertotermostaatit, lämmittävät termostaattihauteet, HBC-sarja
Toimittaja: IKA
4127000 4137000 4125000 4135000
441-0408EA 6250 EUR
441-0408 441-0411 441-0409 441-0410
Kiertotermostaatit, lämmittävät termostaattihauteet, HBC-sarja
Kiertotermostaatit Avoimet termostaattihaudejärjestelmät Hauteet, joissa irrotettavat termostaattihauteet
The HBC circulating baths have a high-quality stainless steel bath, an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphic TFT Display illustrates all relevant process parameters. The large surface of the radiator allows for an optimum heat exchange and a smooth warming of the medium. Units feature push and turn buttons for intuitive menu navigation. All models conform to the highest safety classification III (FL) for use with flammable liquids according to DIN 12876. HBC immersion circulators ensure short heating up times andMore Product Information