Formamidi 99+% spektrofotometriaan

Toimittaja: Thermo Fisher Scientific
Vaara

44066.AE 44066.AP 44066.AY
44066.AEEA 121 EUR
44066.AE 44066.AP 44066.AY
Formamidi 99+% spektrofotometriaan
Formamidi

Purity: 99+% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 154°(310°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Colourless Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941

Kaava: HCONH₂
Kiehumispiste: 210 °C
Sulamispiste: 2 °C
Tiheys: 1,134
Leimahduspiste: 154 °C (310 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007941
CAS-numero: 75-12-7
EINECS: 200-842-0
Merck Index: 14,4237
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR