Magnesium oxalate dihydrate ≥98.5%

Toimittaja: Thermo Fisher Scientific
Varoitus

43976.22 43976.36 43976.A3
43976.22EA 261 EUR
43976.22 43976.36 43976.A3
Magnesium oxalate dihydrate ≥98.5%
Magnesium oxalate dihydrate

Purity: 98.5+% CAS No.: 547-66-0 Molecular Formula: MgC2O4.2H2O Formula Weight: 148.36 (112.32anhy) Physical Form: Powder/Pieces Odour: Odourless Merck Reference: 14,5676 UN No.: 3288 MDL No.: MFCD00287279

Kaava: C₂MgO₄·2H₂O
Sulamispiste: 150 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00287279
CAS-numero: 547-66-0
EINECS: 208-932-1
ADR: 6.1,III
Merck Index: 14,5676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR