Silver standard dissolved in oil, 5.000 µg/g Ag in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

43868.18
43868.18EA 159 EUR
43868.18
Silver standard dissolved in oil, 5.000 µg/g Ag in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Silver Standards

Physical Form: Liquid
MDL No.: MFCD00003397

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR