Arsenic standard dissolved in oil, 1.000 µg/g As in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

43864.1Y 43864.18
43864.1YEA 0 EUR
43864.1Y 43864.18
Arsenic standard dissolved in oil, 1.000 µg/g As in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Arsenic Standards

Physical Form: Liquid
MDL No.: MFCD00085309

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR