Zirconium oxide catalyst support

Toimittaja: Thermo Fisher Scientific

43815.22 43815.A4 43815.36
43815.22EA 206 EUR
43815.22 43815.A4 43815.36
Zirconium oxide catalyst support
Zirconium oxide

Physical Form: 1/8" Pellets Odour: Odourless MDL No.: MFCD00011310


Applications: Catalyst carrier

Kaava: OZr
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011310
CAS-numero: 12036-01-0
EINECS: 215-227-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR