Alumiini ≥99.9995% (metals basis), folio, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)

Toimittaja: Thermo Fisher Scientific

43777.GH 43777.HP
43777.GHEA 325 EUR
43777.GH 43777.HP
Alumiini ≥99.9995% (metals basis), folio, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Alumiini

Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Kaava: Al
Kiehumispiste: 2327 °C (1013 hPa)
Sulamispiste: 660 °C
Tiheys: 2,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134029
CAS-numero: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR