Nikkeli ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Toimittaja: Thermo Fisher Scientific
Vaara

43763.BY 43763.BU 43763.BQ
43763.BYEA 1980 EUR
43763.BY 43763.BU 43763.BQ
Nikkeli ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Nikkeli

Purity: 99.999% CAS No.: 7440-02-0 Physical Form: ≈28.0g/m Odour: Odourless MDL No.: MFCD00011137

Kaava: Ni
MW: 58.69 g/mol
Kiehumispiste: 3007
Sulamispiste: 1453
Tiheys: 8.9 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011137
CAS-numero: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR