Cesiumkloridi, ultra dry ≥99.99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

43720.09 43720.30 43720.18
43720.09EA 292 EUR
43720.09 43720.30 43720.18
Cesiumkloridi, ultra dry ≥99.99% (metals basis)
Cesiumkloridi

Purity: 99.99% CAS No.: 7647-17-8 Molecular Formula: CsCl Formula Weight: 168.36 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.99 Refractive Index: 1.6418 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 646° Boiling Point: 1303° Merck Reference: 14,2011 MDL No.: MFCD00010955

Kaava: CsCl
Kiehumispiste: 1303 °C
Sulamispiste: 646 °C
Tiheys: 3,99
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010955
CAS-numero: 7647-17-8
EINECS: 231-600-2
Merck Index: 14,2011

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR