Piperidiini 99.5%, AcroSeal®, purified by redistillation

Toimittaja: Thermo Fisher Scientific
Vaara

433840010 433841000
433840010.EA 421 EUR
433840010. 433841000.
Piperidiini 99.5%, AcroSeal®, purified by redistillation
Piperidiini
Kaava: C₅H₁₁N
Kiehumispiste: 107,3 °C (1013 hPa)
Sulamispiste: –11 °C
Tiheys: 0,862 g/cm³
Leimahduspiste: 16 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005979
CAS-numero: 110-89-4
UN: 2401
ADR: 8,I

Order Now

Testitulosten erittely

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4540 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR