Rutenium(III)kloridihydraatti ≥99.99% (metals basis), Premion®

Toimittaja: Thermo Fisher Scientific
Vaara

43364.09 43364.04
43364.09EA 1690 EUR
43364.09 43364.04
Rutenium(III)kloridihydraatti ≥99.99% (metals basis), Premion®
Rutenium(III)kloridihydraatti

Purity: 99.99% CAS No.: 14898-67-0 Molecular Formula: RuCl3.xH2O Formula Weight: 207.43(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Melting Point: 100°dec Merck Reference: 14,8302 UN No.: 3260 MDL No.: MFCD00149844

Kaava: Cl₃Ru·xH₂O
Sulamispiste: 100 °C (haj. lämpötila)
Tiheys: 3,11 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149844
CAS-numero: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III
Merck Index: 14,8302

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR