Heptylmagnesium bromide 1 M diethyl ether, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

433031000
433031000.EA 151 EUR
433031000.
Heptylmagnesium bromide 1 M diethyl ether, AcroSeal®
Heptylmagnesium bromide
Kaava: C₇H₁₅BrMg
MW: 203.41 g/mol
Kiehumispiste: 35 °C (760 mmHg)
Tiheys: 0.841 g/cm³
Leimahduspiste: –30 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013517
CAS-numero: 13125-66-1
UN: 2924
ADR: 3 I

Order Now

Testitulosten erittely

Appearance Brown Solution
Titration Grignard 0.95 to 1.10 M
Titration Argentometric 96.0 to 104.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR