Platina ≥99.95% (metals basis), lanka, annealed, Ø 0.5 mm (0.02 in)

Toimittaja: Thermo Fisher Scientific

43288.BY 43288.BU 43288.CC
43288.BYEA 2460 EUR
43288.BY 43288.BU 43288.CC
Platina ≥99.95% (metals basis), lanka, annealed, Ø 0.5 mm (0.02 in)
Platina

Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈4.21g/m, Temper: soft Appearance: Gold Odour: Odourless MDL No.: MFCD00011179

Kaava: Pt
Kiehumispiste: 3827 °C (1013 hPa)
Sulamispiste: 1769 °C
Tiheys: 21,447 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR