Grinding pieces, zirconium oxide, yttria stabilised

Toimittaja: Thermo Fisher Scientific
43286.A1 43285.A1 43286.30 43285.30
43286.A1EA 1120 EUR
43286.A1 43285.A1 43286.30 43285.30
Grinding pieces, zirconium oxide, yttria stabilised
Helmet ja partikkelit Ceramic Beads
The density of the zirconium oxide beads is 6.02 g/cm³

  • MDL No.: MFCD00145546
  • Yttrium oxide (mole) 3%
  • Hardness 12
  • Total impuroty level < 0.1

Storage at ambient temperatures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR