Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%

Toimittaja: Thermo Fisher Scientific
Varoitus

43174.22 43174.03 43174.14 43174.06
43174.22EA 1210 EUR
43174.22 43174.03 43174.14 43174.06
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)]

Purity: 96% CAS No.: 14167-18-1 Molecular Formula: C16H14CoN2O2 Formula Weight: 325.23 Physical Form: Powder Appearance: Black Odour: Odourless MDL No.: MFCD00000009

Kaava: C₁₆H₁₆CoN₂O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000009
CAS-numero: 14167-18-1
EINECS: 238-012-5

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR