4-Metyyli-2-pentanoni 99+% HPLC-analytiikkaan

Toimittaja: Thermo Fisher Scientific
Vaara

43170.K2 43170.K7 43170.M1 43170.M6
43170.K2EA 208 EUR
43170.K2 43170.K7 43170.M1 43170.M6
4-Metyyli-2-pentanoni 99+% HPLC-analytiikkaan
4-Metyyli-2-pentanoni

Purity: 99+% CAS No.: 108-10-1 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 13°(55°F) Physical Form: Liquid Density: 0.801 Refractive Index: 1.3960 Odour: Mild Melting Point: -84° Boiling Point: 117-118° Beilstein Reference: 605399 Merck Reference: 14,5207 UN No.: 1245 MDL No.: MFCD00008938

Kaava: (CH₃)₂CHCH₂COCH₃
Kiehumispiste: 117…118 °C
Sulamispiste: –84 °C
Tiheys: 0,801
Leimahduspiste: 13 °C (55 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008938
CAS-numero: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 14,5207
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Environmentally Preferable Safer Human and Environmental Health
Ympäristösertifikaatti Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR