2-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

431611000
431611000.EA 193 EUR
431611000.
2-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®
2-Bromobenzylmagnesium bromide
Kaava: C₇H₆Br₂Mg
Tiheys: 0,766 g/cm³
Leimahduspiste: –40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01319903
CAS-numero: 56812-60-3
UN: 2924
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear yellow to brown Solution
Titration Grignard 0.15 to 0.35 M
Melting point 84 to 87 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR