Zirconium standard dissolved in oil, 5.000 µg/g Zr in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

43128.1Y 43128.18
43128.1YEA 194 EUR
43128.1Y 43128.18
Zirconium standard dissolved in oil, 5.000 µg/g Zr in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Zirconium Standards

Physical form: liquid
MDL No.: MFCD00011303

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR