Lantaani(III)hydroksidi ≥99.95% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

43123.18 43123.30
43123.18EA 168 EUR
43123.18 43123.30
Lantaani(III)hydroksidi ≥99.95% (REO, rare earth oxide basis)
Lantaani(III)hydroksidi

Purity: 99.95% CAS No.: 14507-19-8 Molecular Formula: La(OH)3 Formula Weight: 189.93 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: dec. MDL No.: MFCD00016160

Kaava: H₃LaO₃
Sulamispiste: Dec.
Tiheys: 4,28 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016160
CAS-numero: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR