VWR® Homogenisers
Toimittaja: VWR Collection
431-0290EA 1860 EUR
431-0290 431-0291 431-0292 431-0293
VWR® Homogenisers
Homogenisaattorit Homogenisaattorit, pöytämalli
These compact homogenisers are suitable for all standard dispersing tasks and intensive mixing. The units are robust, and their stability prevents pulsation oscillations and vibrations, it is also simple and safe to change aggregates for different tasks.
  • Easy operation with an on-off switch and speed control
  • Intelligent variable speed control for long equipment life
  • Low noise level
  • Height adjustable stand (available as an option)
  • Switch aggregate easily
  • Models M, L and XL give speed stability even with changes in viscosity
  • XL model provides protection