Germanium(IV) ethoxide ≥99.995% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

43052.03 43052.14 43052.06
43052.03EA 137 EUR
43052.03 43052.14 43052.06
Germanium(IV) ethoxide ≥99.995% (metals basis)
Germanium(IV) ethoxide

Purity: 99.995% CAS No.: 14165-55-0 Molecular Formula: C8H20GeO4 Formula Weight: 252.84 Flash Point: 49°(121°F) Physical Form: Liquid Density: 1.14 Refractive Index: 1.4070 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Alcohol-like Melting Point: -72° Boiling Point: 185.5° UN No.: 1993 MDL No.: MFCD00015121


Preparation of GeO2 by CVD; intermediate in sol-gel synthesis

Kaava: C₈H₂₀GeO₄
Kiehumispiste: 185,5 °C
Sulamispiste: –72 °C
Tiheys: 1,14
Leimahduspiste: 49 °C (121 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015121
CAS-numero: 14165-55-0
EINECS: 238-007-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR