1-Amino-2-methyl-9,10-anthraquinone, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

42994-25MG
42994-25MGEA 0 EUR
42994-25MG
1-Amino-2-methyl-9,10-anthraquinone, Supelco®
1-Amino-2-methyl-9,10-anthraquinone
Kaava: C₁₅H₁₁NO₂
MDL numero: MFCD00001220
CAS-numero: 82-28-0
EINECS: 201-408-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ac260176a0c631faf9bbc62f6fa825cf858eb77&vid=4939b327875b0ad2dbb32b31efdb228419c0c901&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR