Seleenidisulfidi 94

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Selenium sulphide, Selenium(IV) sulphide

42943.14 42943.22 42943.36
42943.14EA 134 EUR
42943.14 42943.22 42943.36
Seleenidisulfidi 94
Seleenidisulfidi

Purity: 94% CAS No.: 7488-56-4 Molecular Formula: SeS2 Formula Weight: 143.09 Physical Form: Powder Melting Point: 100° Boiling Point: dec. Solubility: Insoluble in water, in dilute HCl Merck Reference: 14,8436 UN No.: 2657 MDL No.: MFCD00011227

Kaava: S₂Se
MW: 143.09 g/mol
Kiehumispiste: -112 Dec.
Tiheys: 0.03 g/cm³ (–65 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011227
CAS-numero: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR