Litiumjodidi, ultra dry ≥99.999% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

42851.04 42851.09 42851.18
42851.04EA 135 EUR
42851.04 42851.09 42851.18
Litiumjodidi, ultra dry ≥99.999% (metals basis)
Litiumjodidi

Purity: 99.999% CAS No.: 10377-51-2 Molecular Formula: LiI Formula Weight: 133.84 Physical Form: Powder, Ampouled under argon Density: 4.076 Refractive Index: 1.955 Sensitivity: Light Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless Melting Point: 449° Boiling Point: 1170-1190° Merck Reference: 14,5535 MDL No.: MFCD00011092

Kaava: ILi
Kiehumispiste: 1170…1190 °C
Sulamispiste: 449 °C
Tiheys: 4,076
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011092
CAS-numero: 10377-51-2
EINECS: 233-822-5
Merck Index: 14,5535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR