1-Octylmagnesium chloride 1.4 M in THF, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

428031000 428038000
428031000.EA 106 EUR
428031000. 428038000.
1-Octylmagnesium chloride 1.4 M in THF, AcroSeal®
1-Octylmagnesium chloride
Kaava: C₈H₁₇ClMg
Leimahduspiste: –10 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000476
CAS-numero: 38841-98-4
UN: 2924
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Grey-green to brown Solution
Titration Grignard 1.2 to 1.6 M
Additional info is possible)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR