n-Octylmagnesium bromide 2 M in diethyl ether, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

427811000 427818000
427811000.EA 86.9 EUR
427811000. 427818000.
n-Octylmagnesium bromide 2 M in diethyl ether, AcroSeal®
n-Octylmagnesium bromide

Appearance: Yellow to brown Solution

Kaava: C₈H₁₇BrMg
Kiehumispiste: 35 °C (760 mmHg)
Tiheys: 0,961 g/cm³
Leimahduspiste: –35 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191566
CAS-numero: 17049-49-9
UN: 2924
ADR: 3,I

Order Now

Testitulosten erittely

Appearance Yellow to brown Solution
Titration Grignard 1.9 to 2.2 M
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR