Silica gel, TLC high purity grade, without binder

Toimittaja: Thermo Fisher Scientific

42756.30 42756.A1 42756.A7
42756.30EA 233 EUR
42756.30 42756.A1 42756.A7
Silica gel, TLC high purity grade, without binder
HPLC- ja TLC, Täytemateriaalit TLC Sorbents

This TLC high purity grade silica gel without binder is used for thin layer chromatography plate preparation.


  • Incompatible with oxidizing agents
  • Soluble in alkalies
  • Insoluble in water
  • pH 6 to 7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR