Magnesium standard dissolved in oil, 5.000 µg/g Mg in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

42443.18
42443.18EA 171 EUR
42443.18
Magnesium standard dissolved in oil, 5.000 µg/g Mg in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Magnesium Standards

Physical Form: Liquid
MDL No.: MFCD00085308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR