Lead standard dissolved in oil, 5.000 µg/g Pb in mineral oil Specpure® for wear metal analysis

Toimittaja: Thermo Fisher Scientific

42441.18
42441.18EA 210 EUR
42441.18
Lead standard dissolved in oil, 5.000 µg/g Pb in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Lead Standards

Physical form: liquid
MDL No.: MFCD00134050

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR