Palladium ≥99.95% (metals basis), sponge , -100/+325 mesh

Toimittaja: Thermo Fisher Scientific
Varoitus

42431.03 42431.06 42431.14
42431.03EA 786 EUR
42431.03 42431.06 42431.14
Palladium ≥99.95% (metals basis), sponge , -100/+325 mesh
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Appearance: Dark grey to black Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167

Kaava: Pd
Kiehumispiste: 3167 °C (1013 hPa)
Sulamispiste: 1555 °C
Tiheys: 12,02 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,III
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR