2-Propanoli 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Isopropyylialkoholi

423830010 423835000 423830025 423830250
423830010.EA 68.6 EUR
423830010. 423835000. 423830025. 423830250.
2-Propanoli 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
2-Propanoli
Kaava: (CH₃)₂CHOH
MW: 60.1 g/mol
Kiehumispiste: 82 °C (1013 hPa)
Sulamispiste: –89.5 °C
Tiheys: 0.785 g/cm³
Leimahduspiste: 12 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011674
CAS-numero: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear liquid
Water ≤0.2 % (K.F.)
Boiling point 81 to 83 °C
Density (20/20 °C) 0.784 to 0.787
Solubility (in water) Passes test
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.002 % (acetone)
Titrable acid or base ≤0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR