Alumiini ≥99.999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Toimittaja: Thermo Fisher Scientific

42324.18 42324.30
42324.18EA 250 EUR
42324.18 42324.30
Alumiini ≥99.999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Alumiini

Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈1.1g/slug Odour: Odourless MDL No.: MFCD00134029

Kaava: Al
Kiehumispiste: 2327 °C (1013 hPa)
Sulamispiste: 660 °C
Tiheys: 2,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134029
CAS-numero: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR