Petrolieetteri, 35…60 °C ACS

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

42085.K2 42085.M1
42085.K2EA 141 EUR
42085.K2 42085.M1
Petrolieetteri, 35…60 °C ACS
Petrolieetteri

CAS No.: 8032-32-4 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.640 Refractive Index: 1.3630 Appearance: Colourless Odour: Petroleum-like Boiling Point: 35-60° UN No.: 1268 MDL No.: MFCD00081849

Kiehumispiste: 35…60 °C
Tiheys: 0.640
Leimahduspiste: –49 °C (–57 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081849
CAS-numero: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR