Urea 98.0-102.0% USP, Multi-Compendial, J.T.Baker®

Toimittaja: Avantor
Varoitus

4203-01EA 208.17 EUR
4203-01 4203-60 4203-07 4203-08
Urea 98.0-102.0% USP, Multi-Compendial, J.T.Baker®
Urea

  • GMP manufactured product


Sertifikaatit: Meets U.S.P requirements and meets B.P., E.P. and J.P. chemical specifications.

Kaava: NH₂CONH₂
Kiehumispiste: 197 °C (1013 hPa)
Tiheys: 1,32 g/ml (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008022
CAS-numero: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets U.S.P Requirements Passes Test
GMP Manufactured Product Passes Test
Meets B.P. Chemical Specifications 98.0 - 102.0%
Meets E.P. Chemical Specifications ≤0.1%
Meets J.P. Chemical Specifications ≤0.04%
CAUTION: For Manufacturing, processing or repackaging ≤0.1%
Bulk Pharmaceutical Chemical ≤0.1%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Residue on Ignition ≤0.1 %
USP - Alcohol Insoluble Matter ≤0.04 %
USP - Organic impurities: Urea related compound A ≤0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤0.1 %
USP - Organic impurities: Total impurities ≤2.0 %
USP - Endotoxin Concentration (EU/g) ≤2.5
EP/BP - Assay (NH₂CONH₂) (dried basis) 98.5 - 101.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Alkalinity Passes Test
EP/BP - Biuret ≤0.1 %
EP/BP - Ammonium (NH₄) ≤500 ppm
EP/BP - Loss on Drying at 105°C ≤1.0 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (NH₂CONH₂) 99.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Chloride (Cl) ≤0.007 %
JP - Sulfate (SO₄) ≤0.010 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Alcohol Insoluble Matter ≤0.040 %
JP - Residue on Ignition ≤0.10 %
JP - Melting Point 132.5 - 134.5 °C
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR