Potassium tellurite hydrate 97%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Potassium tellurite(IV) hydrate, Potassium tellurium trioxide hydrate, di-Potassium tellurium trioxide hydrate, Potassium tellurium (IV) oxide hydrate, Potassium tellurite monohydrate

41959.14 41959.22
41959.14EA 359 EUR
41959.14 41959.22
Potassium tellurite hydrate 97%
Potassium tellurite hydrate

Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.


Purity: 97% CAS No.: 123333-66-4 Molecular Formula: K2O3Te.H2O Formula Weight: 276.81 (253.80anhy) Physical Form: Crystalline Appearance: White Melting Point: 465° dec. UN No.: 3284 MDL No.: MFCD00149923

Kaava: K₂O₃Te·H₂O
MW: 271.81 g/mol
Sulamispiste: 465 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149923
CAS-numero: 123333-66-4
EINECS: 232-213-1
UN: 3284
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR