Magnesium citrate dibasic hydrate

Toimittaja: Thermo Fisher Scientific

41954.30 41954.A1
41954.30EA 278 EUR
41954.30 41954.A1
Magnesium citrate dibasic hydrate
Magnesium citrate dibasic hydrate

CAS No.: 119851-23-9 Molecular Formula: C6H6MgO7.xH2O Formula Weight: 214.14(anhy) Physical Form: Powder Appearance: White to pale yellow MDL No.: MFCD00151706

Kaava: C₆H₆MgO₇·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151706
CAS-numero: 119851-23-9
EINECS: 231-923-9
Merck Index: 14,5663

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR