Strontium tungsten oxide ≥99.9% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

41899.14 41899.22
41899.14EA 221 EUR
41899.14 41899.22
Strontium tungsten oxide ≥99.9% (metals basis)
Strontium tungsten oxide

Purity: 99.9% CAS No.: 13451-05-3 Molecular Formula: O4SrW Formula Weight: 335.46 Physical Form: -200 Mesh Powder Density: 6.187 Appearance: White Odour: Odourless MDL No.: MFCD00049562

Kaava: O₄SrW
Tiheys: 6,187
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00049562
CAS-numero: 13451-05-3
EINECS: 236-617-9

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR