Kalsiumhydroksidi 95.0-100.5% (by EDTA titration), jauhe FCC, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

4188-40EA 6951.05 EUR
4188-40 MACR4188-25 4188-05 4188-24
Kalsiumhydroksidi 95.0-100.5% (by EDTA titration), jauhe FCC, Macron Fine Chemicals™
Kalsiumhydroksidi
Kaava: Ca(OH)₂
Sulamispiste: 550 °C
Tiheys: 2,24 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010901
CAS-numero: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Testitulosten erittely

Food GMP Manufactured Product 95.0 - 100.5%
Meets F.C.C. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
FCC - Assay (Ca(OH)₂) (by EDTA titrn) 95.0 - 100.5 %
Appearance Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Carbonate (CO₃) Passes Test
FCC - Acid-Insoluble Substances ≤0.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤2 ppm
FCC - Fluoride (F) ≤0.005 %
FCC - Magnesium and Alkali Salts ≤4.8 %
Thru U.S. No. 325 Sieve, For Information Only
Barium (Ba), For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR