Poly(styrene sulphonic acid)sodium salt, M.W. 70,000

Toimittaja: Thermo Fisher Scientific

Synonyms: Poly(sodium-p-styrenesulphonate)

41688.18 41688.30
41688.18EA 140 EUR
41688.18 41688.30
Poly(styrene sulphonic acid)sodium salt, M.W. 70,000
Poly(styrene sulphonic acid)sodium salt

CAS No.: 25704-18-1 Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n Physical Form: Powder MDL No.: MFCD00084449

Kaava: [-CH₂CH(C₆H₄)SO₃Na)-]n
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00084449
CAS-numero: 25704-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR