Multi-element QC-21 check standard solution, Specpure®

Toimittaja: Thermo Fisher Scientific

41328.AE 41328.AP
41328.AEEA 1030 EUR
41328.AE 41328.AP
Multi-element QC-21 check standard solution, Specpure®
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards

Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.


Molecular Formula: Matrix: 5% HNO₃/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00803610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR