Kaliumbromidi, ultra dry ≥99.95% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

41282.14 41282.22
41282.14EA 250 EUR
41282.14 41282.22
Kaliumbromidi, ultra dry ≥99.95% (metals basis)
Kaliumbromidi

Purity: 99.95% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Powder, Ampouled under argon Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Kaava: KBr
Kiehumispiste: 1435 °C
Sulamispiste: 730 °C
Tiheys: 2,75
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011358
CAS-numero: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR