Polyvinyylialkoholi, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed

Toimittaja: Thermo Fisher Scientific

41243.14 41243.22 41243.36
41243.14EA 57 EUR
41243.14 41243.22 41243.36
Polyvinyylialkoholi, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed
Polyvinyylialkoholi

Purity: 98-99% CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Physical Form: Solid Odour: Odourless MDL No.: MFCD00081922

Kaava: [-CH₂CH(OH)-]n
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081922
CAS-numero: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR