Nucleic acid stains, PAGE GelRed™
Toimittaja: Biotium
41008-T 41008-500UL 41014
41008-TEA 42.8 EUR
41008-T 41008-500UL 41014.
Nucleic acid stains, PAGE GelRed™
Värit Elektroforeesivärit
Stains specifically designed for DNA in polyacrylamide gels.
  • More sensitive for agarose gel applications
  • Two to three times more sensitive for dsDNA over RNA
  • Impenetrable to latex or nitrile gloves and cell membranes
Unlike the highly mutagenic ethidium bromide (EtBr), PAGE GelRed are non-toxic and non-mutagenic in bacterial AMES tests.
PAGE GelRed can be imaged using a 254 nm UV transilluminator with an ethidium bromide filter.
Sertifikaatit: CCR Title 22 for the management of hazardous waste.