VWR®, Pestles
Toimittaja: VWR Collection
410-0126EA 209 EUR
410-0126 410-0125 410-0124 410-0123 410-0122 410-0121 410-0120 410-0119
VWR®, Pestles
Huhmareet ja survimet Huhmareet
Porcelain.
  • Grinding surface unglazed
  • Glazed handle
Sertifikaatit: Produced according to DIN 12906.