VWR®, Mortars, without Pestles
Toimittaja: VWR Collection
410-0117EA 348 EUR
410-0117 410-0108 410-0118 410-0116 410-0115 410-0114 410-0113 410-0112 410-0111 410-0110 410-0109
VWR®, Mortars, without Pestles
Huhmareet ja survimet Huhmareet
Porcelain, produced according to DIN 12906.
  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis