Natriumlauryylisulfaatti ≥99%, Ultrapure Bioreagent, J.T.Baker®

Toimittaja: Avantor
Vaara

4095-07EA 3812.58 EUR
4095-07 4095-02 4095-04 4095-05
Natriumlauryylisulfaatti ≥99%, Ultrapure Bioreagent, J.T.Baker®
Natriumlauryylisulfaatti

For electrophoresis and protein solubilization.


  • Lot analysis on label

Kaava: CH₃(CH₂)₁₁OSO₃Na
Sulamispiste: 204…207 °C
Tiheys: 0,40 g/cm³
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036175
CAS-numero: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1, 6.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use ≥99%
Appearance Passes Test
Assay (as C₁₂H₂₅OSO₃Na) ≥99 %
Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path) ≤0.1
pH of 1% Solution at 25°C 5.0 - 8.0
Chloride (Cl) ≤0.03 %
Nitrogen (N) ≤5 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR